missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran, Thermo Scientific™

Catalog Number p-4391275
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1131912-76-9
C11H21BO3
212.10
MFCD11506065
NLSMOSUUBUCSPL-UHFFFAOYSA-N
tetrahydropyran-4-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-tetrahydro-2h-pyran-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl tetrahydropyran, tetrahydropyran-4-boronic acid, pinacol ester, tetrahydropyran-4-boronicacidpinacolester, 4,4,5,5-tetramethyl-2-oxan-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl oxane, tetrahydro-2h-pyran-4-yl boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl tetrahydro-2h-pyran
42614649
4,4,5,5-tetramethyl-2-(oxan-4-yl)-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1CCOCC1
This item is not returnable. View return policy
1131912-76-9
C11H21BO3
212.10
MFCD11506065
NLSMOSUUBUCSPL-UHFFFAOYSA-N
tetrahydropyran-4-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-tetrahydro-2h-pyran-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl tetrahydropyran, tetrahydropyran-4-boronic acid, pinacol ester, tetrahydropyran-4-boronicacidpinacolester, 4,4,5,5-tetramethyl-2-oxan-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl oxane, tetrahydro-2h-pyran-4-yl boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl tetrahydro-2h-pyran
42614649
4,4,5,5-tetramethyl-2-(oxan-4-yl)-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1CCOCC1
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 212.09
Percent Purity 97%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'