missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran, Thermo Scientific™

Catalog Number 10505660
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
287944-16-5
C11H19BO3
210.08
MFCD11052631
DOSGEBYQRMBTGS-UHFFFAOYSA-N
3,6-dihydro-2h-pyran-4-boronic acid pinacol ester, 2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester, 3,6-dihydro-2h-pyran-4-boronicacidpinacolester, 3,6-dihydro-2h-pyran-4-boronic acid pinacolester, 3,6 dihydro-2h-pyran-4-boronic acid pinacol ester
11218053
2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CCOCC1
This item is not returnable. View return policy
287944-16-5
C11H19BO3
210.08
MFCD11052631
DOSGEBYQRMBTGS-UHFFFAOYSA-N
3,6-dihydro-2h-pyran-4-boronic acid pinacol ester, 2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester, 3,6-dihydro-2h-pyran-4-boronicacidpinacolester, 3,6-dihydro-2h-pyran-4-boronic acid pinacolester, 3,6 dihydro-2h-pyran-4-boronic acid pinacol ester
11218053
2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CCOCC1
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 210.08
Percent Purity 95%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'