missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4,4,4,4',4',4'-Hexafluoro-DL-valine, 97%

Brand:  Alfa Aesar™ L13072.ME

Code : NEW

Additional Details : CAS Number : 16063-80-2 Weight : 0.01000kg

Quantity 500mg

 View more versions of this product

Product Code. 11367866

  • £104.73 / 500mg


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Specifications

Chemical Identifiers

4,4,4,4',4',4'-Hexafluoro-DL-valine
C5H5F6NO2
MFCD00014349
16063-80-2
225.09
2416649

Specifications

4,4,4,4',4',4'-Hexafluoro-DL-valine
97%
225.09
2416649
500mg
16063-80-2
C5H5F6NO2
MFCD00014349
≈204°C
Videos
SDS