missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4(3H)-Pyrimidone, 98+%, ACROS Organics™
£105.47
Chemical Identifiers
CAS | 51953-17-4 |
---|---|
Molecular Formula | C4H4N2O |
Molecular Weight (g/mol) | 96.09 |
MDL Number | MFCD00167178 |
InChI Key | DNCYBUMDUBHIJZ-UHFFFAOYSA-N |
Synonym | pyrimidin-4-ol, 4-hydroxypyrimidine, 4-pyrimidinol, 4 3h-pyrimidone, pyrimidin-4 3h-one, 4 3h-pyrimidinone, 4-pyrimidone, 4 1h-pyrimidinone, 4-pyrimidinone, deaminoisocytosine |
PubChem CID | 20695 |
IUPAC Name | 1H-pyrimidin-6-one |
SMILES | C1=CN=CNC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10063050
|
Acros Organics
228250100 |
10g | Glass bottle |
£105.47
10g |
In stock, ready to ship: 2 | Please sign in to purchase this item. Need a web account? Register with us today! |
Chemical Identifiers
51953-17-4 | |
96.09 | |
DNCYBUMDUBHIJZ-UHFFFAOYSA-N | |
20695 | |
C1=CN=CNC1=O |
C4H4N2O | |
MFCD00167178 | |
pyrimidin-4-ol, 4-hydroxypyrimidine, 4-pyrimidinol, 4 3h-pyrimidone, pyrimidin-4 3h-one, 4 3h-pyrimidinone, 4-pyrimidone, 4 1h-pyrimidinone, 4-pyrimidinone, deaminoisocytosine | |
1H-pyrimidin-6-one |
Specifications
4(3H)-Pyrimidone | |
Glass bottle | |
163.0°C to 168.0°C | |
98+% | |
98.0 | |
98+% | |
MFCD00167178 | |
pyrimidin-4-ol, 4-hydroxypyrimidine, 4-pyrimidinol, 4 3h-pyrimidone, pyrimidin-4 3h-one, 4 3h-pyrimidinone, 4-pyrimidone, 4 1h-pyrimidinone, 4-pyrimidinone, deaminoisocytosine | |
C1=CN=CNC1=O | |
96.09 | |
96.09 | |
98+% |
Authentic | |
White to Yellow | |
10g | |
51953-17-4 | |
100.0 | |
C4H4N2O | |
24, 81 | |
DNCYBUMDUBHIJZ-UHFFFAOYSA-N | |
1H-pyrimidin-6-one | |
20695 | |
Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4(3H)-Pyrimidone
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.