missing translation for 'onlineSavingsMsg'
Learn More

4(3H)-Pyrimidinone, 98%, Thermo Scientific Chemicals

Catalog Number p-7081014
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
51953-17-4
C4H4N2O
96.089
MFCD00167178
DNCYBUMDUBHIJZ-UHFFFAOYSA-N
pyrimidin-4-ol, 4-hydroxypyrimidine, 4-pyrimidinol, 4 3h-pyrimidone, pyrimidin-4 3h-one, 4 3h-pyrimidinone, 4-pyrimidone, 4 1h-pyrimidinone, 4-pyrimidinone, deaminoisocytosine
20695
1H-pyrimidin-6-one
C1=CN=CNC1=O
This item is not returnable. View return policy
51953-17-4
C4H4N2O
96.089
MFCD00167178
DNCYBUMDUBHIJZ-UHFFFAOYSA-N
pyrimidin-4-ol, 4-hydroxypyrimidine, 4-pyrimidinol, 4 3h-pyrimidone, pyrimidin-4 3h-one, 4 3h-pyrimidinone, 4-pyrimidone, 4 1h-pyrimidinone, 4-pyrimidinone, deaminoisocytosine
20695
1H-pyrimidin-6-one
C1=CN=CNC1=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 164°C to 167°C
Quantity 5 g
Beilstein 106975
Formula Weight 96.09
Percent Purity 98%
Chemical Name or Material 4 (3H)-Pyrimidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'