missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Thienyl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog Number 10652315
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
157730-74-0
C11H8OS
188.244
MFCD04039151
MGPZYZLJJSEFLU-UHFFFAOYSA-N
3280631
4-thiophen-3-ylbenzaldehyde
C1=CC(=CC=C1C=O)C2=CSC=C2
This item is not returnable. View return policy
157730-74-0
C11H8OS
188.244
MFCD04039151
MGPZYZLJJSEFLU-UHFFFAOYSA-N
3280631
4-thiophen-3-ylbenzaldehyde
C1=CC(=CC=C1C=O)C2=CSC=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 4-thiophen-3-yl benzaldehyde, 4-3-thienyl benzaldehyde, 4-3-thienyl benzenecarbaldehyde, 4-thiophen-3-yl-benzaldehyde, benzaldehyde,4-3-thienyl, 4-thien-3-yl benzaldehyde, pubchem15939, thienylbenzenecarbaldehyde, 4-3-thiophenyl benzaldehyde, benzaldehyde, 4-3-thienyl
Formula Weight 188.25
Percent Purity 97%
Chemical Name or Material 4-(3-Thienyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'