missing translation for 'onlineSavingsMsg'
Learn More
Learn More
{4-[3-(Dimethylamino)propoxy]phenyl}methanol, 97%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 1g
Specifications
CAS | 426831-08-5 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C12H19NO2 |
MDL Number | MFCD06202771 |
Quantity | 1 g |
Synonym | 4-3-dimethylamino propoxy phenyl methanol, benzenemethanol,4-3-dimethylamino propoxy, 4-3-dimethylamino propoxy phenyl methan-1-ol |
InChI Key | ZLCIFBOMXUAJPZ-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'