missing translation for 'onlineSavingsMsg'
Learn More

4-{[3-(chloromethyl)isoxazol-5-yl]methyl}morpholine, Tech., Thermo Scientific™

Catalog Number 10683265
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
953408-97-4
C9H13ClN2O2
216.665
MFCD11841078
FZRKHZMWHFBKDV-UHFFFAOYSA-N
4-3-chloromethyl isoxazol-5-yl methyl morpholine, 4-3-chloromethyl-1,2-oxazol-5-yl methyl morpholine, 3-chloromethyl-5-morpholin-4-yl methyl isoxazole
43811054
4-[[3-(chloromethyl)-1,2-oxazol-5-yl]methyl]morpholine
C1COCCN1CC2=CC(=NO2)CCl
This item is not returnable. View return policy
953408-97-4
C9H13ClN2O2
216.665
MFCD11841078
FZRKHZMWHFBKDV-UHFFFAOYSA-N
4-3-chloromethyl isoxazol-5-yl methyl morpholine, 4-3-chloromethyl-1,2-oxazol-5-yl methyl morpholine, 3-chloromethyl-5-morpholin-4-yl methyl isoxazole
43811054
4-[[3-(chloromethyl)-1,2-oxazol-5-yl]methyl]morpholine
C1COCCN1CC2=CC(=NO2)CCl
CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 216.67
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-{[3-(Chloromethyl)isoxazol-5-yl]methyl}morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'