missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-{[3-(chloromethyl)isoxazol-5-yl]methyl}morpholine, Tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 80.0 |
CAS Max % | 100.0 |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 216.67 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 4-{[3-(Chloromethyl)isoxazol-5-yl]methyl}morpholine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'