missing translation for 'onlineSavingsMsg'
Learn More

[4-(3-Bromothien-2-yl)phenyl]methanol, 97%, Thermo Scientific™

Catalog Number 10797495
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
937795-99-8
C11H9BrOS
269.156
MFCD09879972
AQBJKGXCOBEPMC-UHFFFAOYSA-N
4-3-bromothien-2-yl phenyl methanol, 4-3-bromothiophen-2-yl phenyl methanol, 4-3-bromo-2-thienyl phenyl methanol
24229755
[4-(3-bromothiophen-2-yl)phenyl]methanol
C1=CC(=CC=C1CO)C2=C(C=CS2)Br
This item is not returnable. View return policy
937795-99-8
C11H9BrOS
269.156
MFCD09879972
AQBJKGXCOBEPMC-UHFFFAOYSA-N
4-3-bromothien-2-yl phenyl methanol, 4-3-bromothiophen-2-yl phenyl methanol, 4-3-bromo-2-thienyl phenyl methanol
24229755
[4-(3-bromothiophen-2-yl)phenyl]methanol
C1=CC(=CC=C1CO)C2=C(C=CS2)Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 269.16
Percent Purity 97%
Chemical Name or Material [4-(3-Bromothien-2-yl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'