missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(3-Bromo-5-fluorobenzyl)morpholine, 96%

Brand:  Alfa Aesar™ H62673.03

Code : NEW

Additional Details : CAS Number : 281652-25-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449168

  • £121.30 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

4-(3-Bromo-5-fluorobenzyl)morpholine
281652-25-3
274.13
MFCD01307672

Specifications

4-(3-Bromo-5-fluorobenzyl)morpholine
Solid
1g
MFCD01307672
281652-25-3
96%
274.13
Videos
SDS