Alfa Aesar™ 4-(3-Bromo-4-fluorobenzyl)morpholine, 96%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H62455.03

UNSPSC: 12352101

Code: NEW

Additional Details:
Additional Details: CAS Number: 281652-25-3 Weight: 0.00100kg


Quantity 1g

Product Code. 15428968

Quantity Price
1 £ 104.62 / 1g
Estimated Shipment date
from Supplier 16-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-(3-Bromo-4-fluorobenzyl)morpholine
CAS 281652-25-3
Formula Weight 274.13
MDL Number MFCD01307672

Specification

Formula Weight 274.13
Physical Form Solid
Chemical Name or Material 4-(3-Bromo-4-fluorobenzyl)morpholine
Assay 96%
CAS 281652-25-3
Quantity 1g
MDL Number MFCD01307672
Documentation

Technical documents