missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(3-Bromo-2-hydroxyphenyl)-2-(3-pyridyl)thiazole, 97%

Brand:  Alfa Aesar™ H65752.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429876

  • £46.64 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

4-(3-Bromo-2-hydroxyphenyl)-2-(3-pyridyl)thiazole
333.2
MFCD27665342

Specifications

4-(3-Bromo-2-hydroxyphenyl)-2-(3-pyridyl)thiazole
Solid
1g
MFCD27665342
Yellow to orange
97%
333.2
Videos
SDS