missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Bromo-2-fluorobenzyl)morpholine, 96%, Thermo Scientific™

Catalog Number 15408838
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1355247-75-4
C11H13BrFNO
274.13
MFCD21333073
CAIURMIERZMRKZ-UHFFFAOYSA-N
1-bromo-2-fluoro-3-morpholinomethyl benzene, 4-3-bromo-2-fluorophenyl methyl morpholine, acmc-209c2f
70699667
4-[(3-bromo-2-fluorophenyl)methyl]morpholine
FC1=C(CN2CCOCC2)C=CC=C1Br
This item is not returnable. View return policy
1355247-75-4
C11H13BrFNO
274.13
MFCD21333073
CAIURMIERZMRKZ-UHFFFAOYSA-N
1-bromo-2-fluoro-3-morpholinomethyl benzene, 4-3-bromo-2-fluorophenyl methyl morpholine, acmc-209c2f
70699667
4-[(3-bromo-2-fluorophenyl)methyl]morpholine
FC1=C(CN2CCOCC2)C=CC=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 274.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-(3-Bromo-2-fluorobenzyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'