missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 200.24 |
Percent Purity | 97% |
Chemical Name or Material | 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'