missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thienyl)benzenesulfonyl chloride, 95%, Thermo Scientific™

Catalog Number p-4812987
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
209592-91-6
C10H7ClO2S2
258.75
IRDZCMUQNGEQEP-UHFFFAOYSA-N
4-2-thienyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-2-thienyl, 4-thiophen-2-yl benzene-1-sulfonyl chloride, 4-thiophen-2-yl benzenesulfonyl chloride
18387759
4-thiophen-2-ylbenzenesulfonyl chloride
C1=CSC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
This item is not returnable. View return policy
209592-91-6
C10H7ClO2S2
258.75
IRDZCMUQNGEQEP-UHFFFAOYSA-N
4-2-thienyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-2-thienyl, 4-thiophen-2-yl benzene-1-sulfonyl chloride, 4-thiophen-2-yl benzenesulfonyl chloride
18387759
4-thiophen-2-ylbenzenesulfonyl chloride
C1=CSC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
Melting Point 114°C to 115°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 258.75
Percent Purity 95%
Chemical Name or Material 4-(2-Thienyl)benzenesulfonyl chloride
compliance-icons compliance-icons
Product Identifier
  • 4-(2-Thienyl)benzenesulfonyl chloride, 95%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'