missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(2-Thiazolidinyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H62513.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469038

  • £56.92 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

4-(2-Thiazolidinyl)benzeneboronic acid
209.07
MFCD26793811

Specifications

4-(2-Thiazolidinyl)benzeneboronic acid
Solid
209.07
1g
97%
MFCD26793811
Videos
SDS