missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(2-Thiazolidinyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H62513.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15479038

  • £285.14 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

4-(2-Thiazolidinyl)benzeneboronic acid
209.07
MFCD26793811

Specifications

4-(2-Thiazolidinyl)benzeneboronic acid
Solid
209.07
5g
97%
MFCD26793811
Videos
SDS