missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Methyl-1H-imidazol-1-yl)benzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 4-2-methyl-1h-imidazol-1-yl benzonitrile, 4-2-methylimidazol-1-yl benzonitrile, 1-4-cyanophenyl-2-methylimidazole, benzonitrile,4-2-methyl-1h-imidazol-1-yl, benzonitrile, 4-2-methyl-1h-imidazol-1-yl, 4-2-methylimidazolyl benzenecarbonitrile, acmc-1c844 |
Formula Weight | 183.21 |
Percent Purity | 97% |
Chemical Name or Material | 4-(2-Methyl-1H-imidazol-1-yl)benzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'