missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-1H-imidazol-1-yl)aniline, 97%, Thermo Scientific™

Catalog Number p-3541089
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
74852-81-6
C10H11N3
173.219
MFCD06797788
IEZCMVRWKNEHJB-UHFFFAOYSA-N
4-2-methylimidazol-1-yl phenylamine, 4-2-methyl-1h-imidazol-1-yl aniline, 4-2-methyl-imidazol-1-yl-phenylamine, 4-2-methyl-1h-imidazol-1-yl benzenamine, 4-2-methylimidazol-1-yl aniline, 4-2-methyl-1h-imidazol-1-yl phenylamine, benzenamine, 4-2-methyl-1h-imidazol-1-yl, 4-2-methylimidazolyl phenylamine, pubchem19934, 1-4-aminophenyl-2-methylimidazole
3157478
4-(2-methylimidazol-1-yl)aniline
CC1=NC=CN1C2=CC=C(C=C2)N
This item is not returnable. View return policy
74852-81-6
C10H11N3
173.219
MFCD06797788
IEZCMVRWKNEHJB-UHFFFAOYSA-N
4-2-methylimidazol-1-yl phenylamine, 4-2-methyl-1h-imidazol-1-yl aniline, 4-2-methyl-imidazol-1-yl-phenylamine, 4-2-methyl-1h-imidazol-1-yl benzenamine, 4-2-methylimidazol-1-yl aniline, 4-2-methyl-1h-imidazol-1-yl phenylamine, benzenamine, 4-2-methyl-1h-imidazol-1-yl, 4-2-methylimidazolyl phenylamine, pubchem19934, 1-4-aminophenyl-2-methylimidazole
3157478
4-(2-methylimidazol-1-yl)aniline
CC1=NC=CN1C2=CC=C(C=C2)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 4-(2-Methyl-1H-imidazol-1-yl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'