missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Keto-1-benzimidazolinyl)piperidine, 97%, Thermo Scientific Chemicals
138.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 183.0°C to 185.0°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: 3g/L (20°C). Other solubilities: 32g/100mL methanol- 13g/100mL ethanol,7g/100mL 2-propanol- 0.75g/100mL acetone,3.1g/100mL methylene chloride- 9.4g/100mL n,n-dmf |
IUPAC Name | 3-piperidin-4-yl-1H-benzimidazol-2-one |
Formula Weight | 217.27 |
Percent Purity | 97% |
Show More |
Safety and Handling
Product Identifier
- 4-(2-Keto-1-benzimidazolinyl)piperidine
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'