Learn More
Alfa Aesar™ 4'-(2-Iodophenylsulfamoyl)acetanilide, 97%
Brand: Alfa Aesar™ H60656.03
Code : NEW
Additional Details : CAS Number : 19838-05-2 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4'-(2-Iodophenylsulfamoyl)acetanilide | |
C14H13IN2O3S | |
MFCD01213971 |
19838-05-2 | |
416.23 | |
2764345 |
Specifications
4'-(2-Iodophenylsulfamoyl)acetanilide | |
97% | |
416.23 | |
2764345 |
19838-05-2 | |
C14H13IN2O3S | |
MFCD01213971 | |
1g |