missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-(2-Hydroxyhexafluoroisopropyl) benzoic acid is used in chemicals, pharmaceutical research, acts as a pharmaceutical intermediate and reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(2-Hydroxyhexafluoroisopropyl) benzoic acid is used in chemicals, pharmaceutical research, acts as a pharmaceutical intermediate and reagent.
Notes
Incompatible with oxidizing agents.
4-(2-Hydroxyhexafluoroisopropyl) benzoic acid is used in chemicals, pharmaceutical research, acts as a pharmaceutical intermediate and reagent.
Notes
Incompatible with oxidizing agents.
Specifications
Melting Point | 122°C to 126°C |
Odor | Odorless |
Quantity | 1 g |
Beilstein | 2148278 |
Formula Weight | 288.15 |
Percent Purity | 97% |
Chemical Name or Material | 4-(2-Hydroxyhexafluoroisopropyl)benzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'