missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)phenol, 97%, Thermo Scientific™

Catalog Number p-2700563
Click to view available options
Quantity:
5 g
10 g
This item is not returnable. View return policy
501-94-0
C8H10O2
138.166
MFCD00002902
YCCILVSKPBXVIP-UHFFFAOYSA-N
tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol
10393
CHEBI:1879
4-(2-hydroxyethyl)phenol
C1=CC(=CC=C1CCO)O
This item is not returnable. View return policy
501-94-0
C8H10O2
138.166
MFCD00002902
YCCILVSKPBXVIP-UHFFFAOYSA-N
tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol
10393
CHEBI:1879
4-(2-hydroxyethyl)phenol
C1=CC(=CC=C1CCO)O
CAS Min % 97.0
Assay Percent Range 97%
Linear Formula HOC6H4CH2CH2OH
Quantity 5 g
Merck Index 15, 10022
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 4-(2-Hydroxyethyl)phenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'