missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Furyl)aniline, ≥97%, Maybridge

Brand:  Maybridge CC34414DA

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 59147-02-3 Weight : 0.05100kg

Quantity 1g
Packaging Amber glass bottle

 View more versions of this product

Product Code. 10061384

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

4-(2-Furyl)aniline
96.5
97%
C10H9NO
MFCD04039072
59147-02-3
100.0
97%
159.19

Specifications

4-(2-Furyl)aniline
96.5
97%
Amber glass bottle
159.19
1g
59147-02-3
100.0
97%
C10H9NO
MFCD04039072
Documentation

Technical Documents