missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Dimethylamino)ethoxy]benzaldehyde, ≥95%, Maybridge™

£29.13 - £254.00

Chemical Identifiers

CAS 15182-92-0
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD07368978
InChI Key CBOKAZFQZOQTOC-UHFFFAOYSA-N
Synonym 4-2-dimethylamino ethoxy benzaldehyde, p-2-dimethylamino ethoxy benzaldehyde, 4-2-dimethylamino-ethoxy-benzaldehyde, pubchem12557, 4-2-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-ethoxy benzaldehyde, 4-2-dimethylaminoethyloxy benzaldehyde, 4-n,n-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-1-ethoxy benzaldehyde
PubChem CID 84823
IUPAC Name 4-[2-(dimethylamino)ethoxy]benzaldehyde
SMILES CN(C)CCOC1=CC=C(C=C1)C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10011634
SDS View Documents
Maybridge
CC51504CB
250mg
£29.13
250mg
View alternative products
Save Up to 
»
Estimated Shipment
08-01-2021
Please to purchase this item. Need a web account? Register with us today!
10020274
SDS View Documents
Maybridge
CC51504DA
1g
£79.36
1g
View alternative products
Save Up to 
»
Estimated Shipment
08-01-2021
Please to purchase this item. Need a web account? Register with us today!
10356714
SDS View Documents
Maybridge
CC51504DE
5g
£254.00
5g
View alternative products
Save Up to 
»
Estimated Shipment
08-01-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

15182-92-0
193.246
CBOKAZFQZOQTOC-UHFFFAOYSA-N
84823
CN(C)CCOC1=CC=C(C=C1)C=O
C11H15NO2
MFCD07368978
4-2-dimethylamino ethoxy benzaldehyde, p-2-dimethylamino ethoxy benzaldehyde, 4-2-dimethylamino-ethoxy-benzaldehyde, pubchem12557, 4-2-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-ethoxy benzaldehyde, 4-2-dimethylaminoethyloxy benzaldehyde, 4-n,n-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-1-ethoxy benzaldehyde
4-[2-(dimethylamino)ethoxy]benzaldehyde

Specifications

4-[2-(Dimethylamino)ethoxy]benzaldehyde
250mg
94.5
95%
MFCD07368978
CBOKAZFQZOQTOC-UHFFFAOYSA-N
4-[2-(dimethylamino)ethoxy]benzaldehyde
84823
95%
Amber Glass Bottle
15182-92-0
100.0
C11H15NO2
4-2-dimethylamino ethoxy benzaldehyde, p-2-dimethylamino ethoxy benzaldehyde, 4-2-dimethylamino-ethoxy-benzaldehyde, pubchem12557, 4-2-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-ethoxy benzaldehyde, 4-2-dimethylaminoethyloxy benzaldehyde, 4-n,n-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-1-ethoxy benzaldehyde
CN(C)CCOC1=CC=C(C=C1)C=O
193.246
193.24
SDS