missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Chloromethyl)-4-(trifluoromethyl)phenyl]morpholine, 97%, Thermo Scientific™

Catalog Number p-3522520
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
892502-15-7
C12H13ClF3NO
279.687
MFCD09025889
KCGVSWOLBMMCMW-UHFFFAOYSA-N
4-2-chloromethyl-4-trifluoromethyl phenyl morpholine, morpholine,4-2-chloromethyl-4-trifluoromethyl phenyl
18525887
4-[2-(chloromethyl)-4-(trifluoromethyl)phenyl]morpholine
C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)CCl
This item is not returnable. View return policy
892502-15-7
C12H13ClF3NO
279.687
MFCD09025889
KCGVSWOLBMMCMW-UHFFFAOYSA-N
4-2-chloromethyl-4-trifluoromethyl phenyl morpholine, morpholine,4-2-chloromethyl-4-trifluoromethyl phenyl
18525887
4-[2-(chloromethyl)-4-(trifluoromethyl)phenyl]morpholine
C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)CCl
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 279.69
Percent Purity 97%
Chemical Name or Material 4-[2-(Chloromethyl)-4-(trifluoromethyl)phenyl]morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'