missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloro-6-nitrophenyl)morpholine, 97%, Thermo Scientific™

Catalog Number 15400599
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
65152-10-5
This item is not returnable. View return policy
65152-10-5

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 242.66
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(2-Chloro-6-nitrophenyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'