missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Bromoethyl)-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™

Catalog Number p-3075525
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
83467-28-1
C7H11BrN2
203.08
MFCD00480987
HGGLMDBNLJUEKA-UHFFFAOYSA-N
4-2-bromoethyl-3,5-dimethyl-1h-pyrazole, 4-2-bromo-ethyl-3,5-dimethyl-1h-pyrazole, 3,5-dimethyl-4-2-bromoethyl-1h-pyrazole, 4-2-bromoethyl-3,5-dimethyl-1h-pyrazol, 1h-pyrazole, 4-2-bromoethyl-3,5-dimethyl, 4-2-bromoethyl-3,5-dimethylpyrazole, akos pao-0750, pubchem12761, timtec-bb sbb010155, aronis23735
572879
4-(2-bromoethyl)-3,5-dimethyl-1H-pyrazole
CC1=C(CCBr)C(C)=NN1
This item is not returnable. View return policy
83467-28-1
C7H11BrN2
203.08
MFCD00480987
HGGLMDBNLJUEKA-UHFFFAOYSA-N
4-2-bromoethyl-3,5-dimethyl-1h-pyrazole, 4-2-bromo-ethyl-3,5-dimethyl-1h-pyrazole, 3,5-dimethyl-4-2-bromoethyl-1h-pyrazole, 4-2-bromoethyl-3,5-dimethyl-1h-pyrazol, 1h-pyrazole, 4-2-bromoethyl-3,5-dimethyl, 4-2-bromoethyl-3,5-dimethylpyrazole, akos pao-0750, pubchem12761, timtec-bb sbb010155, aronis23735
572879
4-(2-bromoethyl)-3,5-dimethyl-1H-pyrazole
CC1=C(CCBr)C(C)=NN1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 203.08
Percent Purity 97%
Chemical Name or Material 4-(2-Bromoethyl)-3, 5-dimethyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'