missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEBSF Hydrochloride, 4-(2-Aminoethyl)-benzenesulfonyl Fluoride HCl, White Powder, Fisher BioReagents
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
1 g
Packaging:
Amber Glass
Description
- Irreverisble serine protease inhibitor
- Inhibits proteases like trypsin, chymotrypsin, kallikrein, plasmin, and thrombin
- Acts as Sulfonyl fluoride that reacts with hydroxy group of the active site serine residue
- More soluble and less toxic
Specifications
Melting Point | 175°C |
Color | White |
Infrared Spectrum | Pass Test |
Assay Percent Range | ≥97 % |
Packaging | Amber Glass |
Quantity | 1 g |
Formula Weight | 239.7 |
Percent Purity | 97% min. |
Physical Form | Powder or Solid |
Chemical Name or Material | 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'