missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Tetrazol-5-yl)benzyl alcohol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Specifications
Quantity | 250 mg |
Formula Weight | 176.18 |
Percent Purity | 97% |
Chemical Name or Material | 4-(1H-Tetrazol-5-yl)benzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'