missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrrol-1-yl)benzylamine, 97%, Maybridge

Brand:  Maybridge CC25513DA

Code : 31

Additional Details : CAS Number : 465514-27-6 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10324242

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

4-(1H-pyrrol-1-yl)benzylamine
96.5
97%
C11H12N2
MFCD03659728
465514-27-6
100.0
97%
172.23

Specifications

4-(1H-pyrrol-1-yl)benzylamine
465514-27-6
100.0
97%
C11H12N2
MFCD03659728
1g
96.5
Amber Glass Bottle
97%
172.23
Videos
SDS