missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrrol-1-yl)aniline, 97%, Thermo Scientific™

Catalog Number p-2700470
Click to view available options
Quantity:
250 mg
1 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
52768-17-9
C10H10N2
158.204
MFCD01934575
NHLHWHRXMZZWGA-UHFFFAOYSA-N
4-1h-pyrrol-1-yl aniline, 4-1-pyrrolyl aniline, 4-pyrrol-1-yl aniline, benzenamine, 4-1h-pyrrol-1-yl, 4-pyrrol-1-yl-phenylamine, 4-pyrrolylphenylamine, 1-p-amino-phenylazole, 4-pyrrol-1-yl-aniline, 1-4-aminophenyl pyrrole, 1-4-amino-phenyl pyrrole
2795457
4-pyrrol-1-ylaniline
C1=CN(C=C1)C2=CC=C(C=C2)N
This item is not returnable. View return policy
52768-17-9
C10H10N2
158.204
MFCD01934575
NHLHWHRXMZZWGA-UHFFFAOYSA-N
4-1h-pyrrol-1-yl aniline, 4-1-pyrrolyl aniline, 4-pyrrol-1-yl aniline, benzenamine, 4-1h-pyrrol-1-yl, 4-pyrrol-1-yl-phenylamine, 4-pyrrolylphenylamine, 1-p-amino-phenylazole, 4-pyrrol-1-yl-aniline, 1-4-aminophenyl pyrrole, 1-4-amino-phenyl pyrrole
2795457
4-pyrrol-1-ylaniline
C1=CN(C=C1)C2=CC=C(C=C2)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrrol-1-yl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'