missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Pyrazol-1-ylmethyl)benzylamine hydrochloride , Tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 80.0 |
CAS Max % | 100.0 |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 223.71 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 4-(1H-pyrazol-1-ylmethyl)benzylamine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'