missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Pyrazol-1-yl)benzaldehyde, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
25 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Quantity | 25 g |
Formula Weight | 172.19 |
Percent Purity | 95% |
Chemical Name or Material | 4-(1H-Pyrazol-1-yl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'