missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Imidazol-1-ylmethyl)-N-methylbenzylamine 1.5 oxalate 0.5 hydrate, 97%, Thermo Scientific™

Catalog Number p-3522501
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
910036-84-9
C14H17N3O4
291.307
MFCD09064952
AIQAEMDKVTYASZ-UHFFFAOYSA-N
oxalic acid; 4-imidazol-1-ylmethyl phenyl methyl methyl amine, oxalic acid-1-4-1h-imidazol-1-yl methyl phenyl-n-methylmethanamine 1/1
24229491
1-[4-(imidazol-1-ylmethyl)phenyl]-N-methylmethanamine;oxalic acid
CNCC1=CC=C(C=C1)CN2C=CN=C2.C(=O)(C(=O)O)O
This item is not returnable. View return policy
910036-84-9
C14H17N3O4
291.307
MFCD09064952
AIQAEMDKVTYASZ-UHFFFAOYSA-N
oxalic acid; 4-imidazol-1-ylmethyl phenyl methyl methyl amine, oxalic acid-1-4-1h-imidazol-1-yl methyl phenyl-n-methylmethanamine 1/1
24229491
1-[4-(imidazol-1-ylmethyl)phenyl]-N-methylmethanamine;oxalic acid
CNCC1=CC=C(C=C1)CN2C=CN=C2.C(=O)(C(=O)O)O
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 345.34
Percent Purity 95%
Chemical Name or Material 4-(1H-imidazol-1-ylmethyl)-N-methylbenzylamine 1.5 oxalate 0.5 hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'