missing translation for 'onlineSavingsMsg'
Learn More

Maybridge 4-(1H-Imidazol-1-ylmethyl)benzonitrile, 10g, 97%

Brand:  Maybridge CC43316EA

4-(1H-Imidazol-1-ylmethyl)benzonitrile, 10g, 97%

Code : 31

Additional Details : CAS Number : 112809-54-8 Weight : 0.06000kg

Product Code. 10672675

  • £246.00 / 10g


Estimated Shipment date
from Supplier 29-05-2018
Add to basket