missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[4-(1H-Imidazol-1-yl)phenyl]methanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 174.2 |
Percent Purity | 97% |
Chemical Name or Material | [4-(1H-Imidazol-1-yl)phenyl]methanol |
Safety and Handling
Product Identifier
- [4-(1H-Imidazol-1-yl)phenyl]methanol
- Warning
- Acute toxicity Category 4
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : [4-(1H-Imidazol-1-yl)phenyl]methanol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'