Learn More
Thermo Scientific Maybridge 4-(1H-Imidazol-1-yl)benzoic acid, 10g, 90%
Brand: Thermo Scientific Maybridge CC18501EA
4-(1H-Imidazol-1-yl)benzoic acid, 10g, 90%
Code : 31
Additional Details : CAS Number : 17616-04-5 Weight : 0.01000kg
Brand: Thermo Scientific Maybridge CC18501EA
4-(1H-Imidazol-1-yl)benzoic acid, 10g, 90%
Code : 31
Additional Details : CAS Number : 17616-04-5 Weight : 0.01000kg