missing translation for 'onlineSavingsMsg'
Learn More

[4-(1H-1,2,4-Triazol-1-ylmethyl)phenyl]methanol, ≥97%, Thermo Scientific™

Catalog Number p-3176185
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
160388-56-7
C10H11N3O
189.218
MFCD07772869
XEOWXPICRQBFIT-UHFFFAOYSA-N
4-1h-1,2,4-triazol-1-ylmethyl phenyl methanol, 4-1,2,4-triazol-1-ylmethyl phenyl methanol, 4-1h-1,2,4-triazol-1-ylmethyl benzyl alcohol, 4-1,2,4-triazolylmethyl phenyl methan-1-ol, 4-1h-1,2,4-triazol-1-yl methyl phenyl methanol
7162035
[4-(1,2,4-triazol-1-ylmethyl)phenyl]methanol
C1=CC(=CC=C1CN2C=NC=N2)CO
This item is not returnable. View return policy
160388-56-7
C10H11N3O
189.218
MFCD07772869
XEOWXPICRQBFIT-UHFFFAOYSA-N
4-1h-1,2,4-triazol-1-ylmethyl phenyl methanol, 4-1,2,4-triazol-1-ylmethyl phenyl methanol, 4-1h-1,2,4-triazol-1-ylmethyl benzyl alcohol, 4-1,2,4-triazolylmethyl phenyl methan-1-ol, 4-1h-1,2,4-triazol-1-yl methyl phenyl methanol
7162035
[4-(1,2,4-triazol-1-ylmethyl)phenyl]methanol
C1=CC(=CC=C1CN2C=NC=N2)CO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.22
Percent Purity 97%
Chemical Name or Material [4-(1H-1, 2, 4-triazol-1-ylmethyl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'