missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-1,2,4-triazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Synonym | 4-1h-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1h-1,2,4-triazol-1-yl methyl benzoic acid, 4-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1,2,4 triazol-1-ylmethyl-benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl benzoic acid hydrochloride, 4-1,2,4-triazolylmethyl benzoic acid, 4-1h-1,2,4-triazole-1-ylmethyl benzoic acid, benzoic acid,4-1h-1,2,4-triazol-1-ylmethyl, benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl |
Formula Weight | 203.2 |
Percent Purity | 97% |
Chemical Name or Material | 4-(1H-1, 2, 4-triazol-1-ylmethyl)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'