missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-Triazol-1-ylmethyl)benzaldehyde 97+%, Maybridge

Brand:  Maybridge CC45704DE

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 859850-94-5 Weight : 0.05500kg Transport : UN number : 1325 Chem class : 1 Pack group : III

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10683635

  • £452.00 / 5g


Estimated Shipment date
from Supplier 04-09-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

4-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
97%
187.2
97%
C10H9N3O

Specifications

4-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
Amber Glass Bottle
97%
187.2
859850-94-5
97%
C10H9N3O
5g
Documentation

Technical Documents