missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-Triazol-1-ylmethyl)benzaldehyde 97+%, Thermo Scientific™

Catalog Number p-4510681
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
859850-94-5
C10H9N3O
187.202
XEPIORKIFKJATD-UHFFFAOYSA-N
4-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazolylmethyl benzaldehyde, 4-1h-1,2,4-triazol-1-yl methyl benzaldehyde, benzaldehyde,4-1h-1,2,4-triazol-1-ylmethyl
7164655
4-(1,2,4-triazol-1-ylmethyl)benzaldehyde
C1=CC(=CC=C1CN2C=NC=N2)C=O
This item is not returnable. View return policy
859850-94-5
C10H9N3O
187.202
XEPIORKIFKJATD-UHFFFAOYSA-N
4-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazolylmethyl benzaldehyde, 4-1h-1,2,4-triazol-1-yl methyl benzaldehyde, benzaldehyde,4-1h-1,2,4-triazol-1-ylmethyl
7164655
4-(1,2,4-triazol-1-ylmethyl)benzaldehyde
C1=CC(=CC=C1CN2C=NC=N2)C=O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'