Learn More
Thermo Scientific Maybridge 4-(1H-1,2,4-Triazol-1-yl)phenol, 10g, 97%
Brand: Thermo Scientific Maybridge AC18635EA
4-(1H-1,2,4-Triazol-1-yl)phenol, 10g, 97%
Code : 31
Additional Details : CAS Number : 68337-15-5 Weight : 0.06000kg
Brand: Thermo Scientific Maybridge AC18635EA
4-(1H-1,2,4-Triazol-1-yl)phenol, 10g, 97%
Code : 31
Additional Details : CAS Number : 68337-15-5 Weight : 0.06000kg