missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-1,2,4-Triazol-1-yl)benzoic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 4-1h-1,2,4-triazol-1-yl benzoic acid, 4-1,2,4 triazol-1-yl-benzoic acid, 4-1,2,4-triazol-1-yl benzoic acid, benzoic acid, 4-1h-1,2,4-triazol-1-yl, benzoic acid,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzoic acid, pubchem19582, rarechem al be 1325, buttpark 9550-80, acmc-1c5ri |
Formula Weight | 189.17 |
Percent Purity | 95% |
Chemical Name or Material | 4-(1H-1, 2, 4-Triazol-1-yl)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'