missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-1,2,4-Triazol-1-yl)benzaldehyde, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 4-1h-1,2,4-triazol-1-yl benzaldehyde, 4-1,2,4 triazol-1-yl-benzaldehyde, 4-1,2,4-triazol-1-yl benzaldehyde, 4-1h-1,2,4-triazol-1-yl benzenecarbaldehyde, benzaldehyde, 4-1h-1,2,4-triazol-1-yl, 1-4-formylphenyl-1,2,4-triazole, benzaldehyde,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzaldehyde, 4-1,2,4-triazol-1-yl-benzaldehyde |
Formula Weight | 173.17 |
Percent Purity | 95% |
Chemical Name or Material | 4-(1H-1, 2, 4-Triazol-1-yl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'