missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-triazol-1-yl)aniline, 97%, Thermo Scientific™

Catalog Number 10724193
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
6523-49-5
C8H8N4
160.18
MFCD00171470
BLNPEJXSNSBBNM-UHFFFAOYSA-N
4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
735826
4-(1,2,4-triazol-1-yl)aniline
C1=CC(=CC=C1N)N2C=NC=N2
This item is not returnable. View return policy
6523-49-5
C8H8N4
160.18
MFCD00171470
BLNPEJXSNSBBNM-UHFFFAOYSA-N
4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
735826
4-(1,2,4-triazol-1-yl)aniline
C1=CC(=CC=C1N)N2C=NC=N2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'