missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse 4-1BB Ligand/TNFSF9 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | 4-1BB Ligand/TNFSF9 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
4-1BB Ligand/TNFSF9 Polyclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot.Specifications
4-1BB Ligand/TNFSF9 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
4-1BB ligand, 41BB Ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, TNFSF9, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
TNFSF9 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P41274 | |
8744 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse 4-1BB Ligand/TNFSF9 Arg104-Glu309 Accession # P41274 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse 4-1BB Ligand/TNFSF9 in direct ELISAs and Western blots. In Western blots, approximatley 5% cross-reactivity with recombinant mouse (rm) Fas Ligand is observed and less than 1% cross-reactivity with rmOX40 Ligand, rmTWEAK, rmTRAIL, and rmTNF-alpha is observed. |
For Research Use Only