missing translation for 'onlineSavingsMsg'
Learn More

4-{[1-(tert-butoxycarbonyl)piperid-4-yloxy]methyl}benzoic acid, 97%, Thermo Scientific™

Catalog Number p-3880862
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
934570-52-2
C18H25NO5
335.4
MFCD11841069
CGLNCSVUPUKDKT-UHFFFAOYSA-N
4-1-tert-butoxycarbonyl piperidin-4-yl oxy methyl benzoic acid, 4-1-tert-butoxycarbonyl piperid-4-yloxy methyl benzoic acid, 4-4-carboxybenzyl oxy piperidine, n-boc protected
43811046
4-[[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxymethyl]benzoic acid
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)C(=O)O
This item is not returnable. View return policy
934570-52-2
C18H25NO5
335.4
MFCD11841069
CGLNCSVUPUKDKT-UHFFFAOYSA-N
4-1-tert-butoxycarbonyl piperidin-4-yl oxy methyl benzoic acid, 4-1-tert-butoxycarbonyl piperid-4-yloxy methyl benzoic acid, 4-4-carboxybenzyl oxy piperidine, n-boc protected
43811046
4-[[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxymethyl]benzoic acid
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 335.4
Percent Purity 97%
Chemical Name or Material 4-{[1-(Tert-butoxycarbonyl)piperid-4-yloxy]methyl}benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'