missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-{[1-(tert-Butoxycarbonyl)-4-piperidinyl]oxy}benzoic acid, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 321.37 |
Percent Purity | 97% |
Chemical Name or Material | 4-{[1-(tert-Butoxycarbonyl)-4-piperidinyl]oxy}benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'